Learn More
Apexbio Technology LLC Vitamin C(Synonyms: Ascorbic Acid, L-Ascorbic Acid, Ascorbate, L-(+)-Ascorbic Acid), 1g, CAS: 50-81-7.
SDP
Catalog No. 5000060551
Encompass_Preferred
Vitamin C (CAS 50-81-7) is a water-soluble vitamin studied extensively in biomedical contexts for its potential anticancer and antiviral activities Mechanistically vitamin C exerts antiproliferative effects on tumor cells and promotes apoptosis as demonstrated in murine colon cancer (CT26) cells At concentrations of 100-200 g/ml vitamin C inhibited proliferation significantly higher levels (200-1000 g/ml) induced apoptosis dose-dependently In vivo studies showed vitamin C treatment reduced tumor volume markedly in CT26 and 4T1 tumor-bearing BALB/c mouse models These findings highlight vitamin C s utility in cancer research as a therapeutic adjunct and investigational agent
Catalog No. 50-000-60551 Supplier Apexbio Technology LLC Supplier No. B20641000
Vitamin C (CAS 50-81-7) is a water-soluble vitamin studied extensively in biomedical contexts for its potential anticancer and antiviral activities Mechanistically vitamin C exerts antiproliferative effects on tumor cells and promotes apoptosis as demonstrated in murine colon cancer (CT26) cells At concentrations of 100-200 g/ml vitamin C inhibited proliferation significantly higher levels (200-1000 g/ml) induced apoptosis dose-dependently In vivo studies showed vitamin C treatment reduced tumor volume markedly in CT26 and 4T1 tumor-bearing BALB/c mouse models These findings highlight vitamin C s utility in cancer research as a therapeutic adjunct and investigational agent