Learn More
Cayman Chemical VItamIn D AChE Tracer 100dtn
SDP
Catalog No. 502945798
Encompass_Preferred
13-HODE cholesteryl ester was originally extracted from atherosclerotic lesions{2227} and shown to be produced by Cu2 -catalyzed oxidation of LDL.{2228} Later studies determined that 15-LO from rabbit reticulocytes and human monocytes were able to metabolize cholesteryl linoleate a major component of LDL to 13-HODE cholesteryl ester.{11262412}
Catalog No. 50-294-5798 Supplier Cayman Chemical Supplier No. 401050100 DTN
13-HODE cholesteryl ester was originally extracted from atherosclerotic lesions{2227} and shown to be produced by Cu2 -catalyzed oxidation of LDL.{2228} Later studies determined that 15-LO from rabbit reticulocytes and human monocytes were able to metabolize cholesteryl linoleate a major component of LDL to 13-HODE cholesteryl ester.{11262412}