Learn More
Athens Research And Technology Inc VITAMIN D BINDING PROTEIN 1MG
Supplier:  Athens Research And Technology Inc 1616070307(VDBP)1MG
Encompass_Preferred
Vitamin D Binding Protein (GC Globulin): 1 MG; Lyophilized from 20 mM Na phosphate, pH 7.4, with 150 mM NaCl; MW 56,000; Purity:> =95% by SDS-PAGE. Native; Prepared and purified from human plasma in the Athens Research & Tech lab.
Catalog No. 50-489-941