Learn More
ABclonal Technology Vitamin D Binding protein
SDP
Catalog No. 5000007900
Encompass_Preferred
The protein encoded by this gene belongs to the albumin gene family It is a multifunctional protein found in plasma ascitic fluid cerebrospinal fluid and on the surface of many cell types It binds to vitamin D and its plasma metabolites and transports them to target tissues Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-07900 Supplier ABclonal Technology Supplier No. A21977100UL
The protein encoded by this gene belongs to the albumin gene family It is a multifunctional protein found in plasma ascitic fluid cerebrospinal fluid and on the surface of many cell types It binds to vitamin D and its plasma metabolites and transports them to target tissues Alternatively spliced transcript variants encoding different isoforms have been found for this gene