Learn More
Apexbio Technology LLC vitamin D binding protein precursor (353-363) [Homo sapiens] 10mg
SDP
Catalog No. 5000096705
Encompass_Preferred
Vitamin D binding protein precursor 353-363 is a serum glycoprotein fragment derived from Homo sapiens It is associated with vitamin D binding protein (DBP) a plasma protein that binds vitamin D metabolites and actin monomers mediating transport in the bloodstream and regulation of extracellular actin levels DBP is present on surfaces of various immune cell types including B and T lymphocytes as well as placental cytotrophoblasts Based on these pharmacological properties vitamin D binding protein precursor 353-363 holds research potential in studies of vitamin D metabolism immune regulation and cytoskeletal interactions
Catalog No. 50-000-96705 Supplier Apexbio Technology LLC Supplier No. A109910
Vitamin D binding protein precursor 353-363 is a serum glycoprotein fragment derived from Homo sapiens It is associated with vitamin D binding protein (DBP) a plasma protein that binds vitamin D metabolites and actin monomers mediating transport in the bloodstream and regulation of extracellular actin levels DBP is present on surfaces of various immune cell types including B and T lymphocytes as well as placental cytotrophoblasts Based on these pharmacological properties vitamin D binding protein precursor 353-363 holds research potential in studies of vitamin D metabolism immune regulation and cytoskeletal interactions