Learn More
MOLECULAR DEPOT LLC Vitamin D BSA - 0.5 mg
Catalog No. 5000013581
Encompass_Preferred
Vitamin D BSA Conjugate Solution (0 5 mg) is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to 25-OH Vitamin D3 A carbon spacer arm links BSA to the 25-OH Vitamin D3 moiety Each BSA molecule contains several 25-OH Vitamin D3 units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline For in vitro research use only
Catalog No. 50-000-13581 Supplier MOLECULAR DEPOT LLC Supplier No. P2010018
Vitamin D BSA Conjugate Solution (0 5 mg) is a concentrated solution of Bovine Serum Albumin (BSA) conjugated to 25-OH Vitamin D3 A carbon spacer arm links BSA to the 25-OH Vitamin D3 moiety Each BSA molecule contains several 25-OH Vitamin D3 units Dilutions of the provided conjugate can be made in Phosphate Buffer Saline For in vitro research use only