Learn More
MOLECULAR DEPOT LLC Vitamin D Depleted - 50 mL
Catalog No. 5000013469
Encompass_Preferred
Vitamin D Depleted Serum (50 mL) is human serum containing undetectable levels (less than 2 ng/mL) of 25-OH vitamin D (either 25-OH vitamin D2 or 25-OH vitamin D3) This product is useful for the dilution of 25-OH vitamin D solutions as well as the research and development of 25-OH vitamin D high and low working solutions This product is liquid stable For in vitro research use only
Catalog No. 50-000-13469 Supplier MOLECULAR DEPOT LLC Supplier No. S2010003
Vitamin D Depleted Serum (50 mL) is human serum containing undetectable levels (less than 2 ng/mL) of 25-OH vitamin D (either 25-OH vitamin D2 or 25-OH vitamin D3) This product is useful for the dilution of 25-OH vitamin D solutions as well as the research and development of 25-OH vitamin D high and low working solutions This product is liquid stable For in vitro research use only