Learn More
MOLECULAR DEPOT LLC Vitamin D Magnetic Bead - 1 mL
Catalog No. 5000013549
Encompass_Preferred
25-OH Vitamin D3 Magnetic Beads consists of a suspension of magnetic particles (1-3 um 0 1perc solid) conjugated to 25-OH vitamin D3 These particles can be recovered easily using a magnet allowing for various applications such as purification depletion assay development and more For in vitro research use only
Catalog No. 50-000-13549 Supplier MOLECULAR DEPOT LLC Supplier No. B2010613
25-OH Vitamin D3 Magnetic Beads consists of a suspension of magnetic particles (1-3 um 0 1perc solid) conjugated to 25-OH vitamin D3 These particles can be recovered easily using a magnet allowing for various applications such as purification depletion assay development and more For in vitro research use only