Learn More
Medchemexpress LLC Vitamin D Receptor/V 100ug
SDP
Catalog No. 5000180128
Encompass_Preferred
Vitamin D receptor (VDR) protein is the nuclear receptor of calcitriol which translocates to the nucleus after binding vitamin D3 thereby coordinating cellular activities VDR forms a heterodimer with RXR binds to DNA response elements and initiates the transcription of vitamin D3-responsive genes Vitamin D Receptor/VDR Protein Mouse (sf9 His) is the recombinant mouse-derived Vitamin D Receptor/VDR protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-80128 Supplier Medchemexpress LLC Supplier No. HYP73542100UG
Vitamin D receptor (VDR) protein is the nuclear receptor of calcitriol which translocates to the nucleus after binding vitamin D3 thereby coordinating cellular activities VDR forms a heterodimer with RXR binds to DNA response elements and initiates the transcription of vitamin D3-responsive genes Vitamin D Receptor/VDR Protein Mouse (sf9 His) is the recombinant mouse-derived Vitamin D Receptor/VDR protein expressed by Sf9 insect cells with C-His labeled tag