Learn More
Medchemexpress LLC Vitamin D Receptor/V 50ug
SDP
Catalog No. 5000180053
Encompass_Preferred
Vitamin D receptor (VDR) protein is the nuclear receptor of calcitriol which translocates to the nucleus after binding vitamin D3 thereby coordinating cellular activities VDR forms a heterodimer with RXR binds to DNA response elements and initiates the transcription of vitamin D3-responsive genes Vitamin D Receptor/VDR Protein Mouse (sf9 His) is the recombinant mouse-derived Vitamin D Receptor/VDR protein expressed by Sf9 insect cells with C-His labeled tag
Catalog No. 50-001-80053 Supplier Medchemexpress LLC Supplier No. HYP7354250UG
Vitamin D receptor (VDR) protein is the nuclear receptor of calcitriol which translocates to the nucleus after binding vitamin D3 thereby coordinating cellular activities VDR forms a heterodimer with RXR binds to DNA response elements and initiates the transcription of vitamin D3-responsive genes Vitamin D Receptor/VDR Protein Mouse (sf9 His) is the recombinant mouse-derived Vitamin D Receptor/VDR protein expressed by Sf9 insect cells with C-His labeled tag