Learn More
Medchemexpress LLC Vitamin D Receptor/V 50ug
SDP
Catalog No. 5000180751
Encompass_Preferred
Vitamin D receptor (VDR) acts as a nuclear receptor for calcitriol the active form of vitamin D3 triggering cellular effects After vitamin D3 binding VDR translocates to the nucleus and forms a heterodimer with the retinoid X receptor (RXR) Vitamin D Receptor/VDR Protein Human (sf9 His) is the recombinant human-derived Vitamin D Receptor/VDR protein expressed by Sf9 insect cells with C-10 His labeled tag
Catalog No. 50-001-80751 Supplier Medchemexpress LLC Supplier No. HYP7354350UG
Vitamin D receptor (VDR) acts as a nuclear receptor for calcitriol the active form of vitamin D3 triggering cellular effects After vitamin D3 binding VDR translocates to the nucleus and forms a heterodimer with the retinoid X receptor (RXR) Vitamin D Receptor/VDR Protein Human (sf9 His) is the recombinant human-derived Vitamin D Receptor/VDR protein expressed by Sf9 insect cells with C-10 His labeled tag