Learn More
MOLECULAR DEPOT LLC Vitamin D Release - 50 mL
Catalog No. 5000013482
Encompass_Preferred
Vitamin D Release Buffer (50 mL) is an aqueous buffer designed to the fast and efficient release vitamin D and its metabolites from their serum/plasma transporter (vitamin D binding protein) Unlike other release agents this buffer is organic solvent-free and liquid stable For in vitro research use only
Catalog No. 50-000-13482 Supplier MOLECULAR DEPOT LLC Supplier No. B2010548
Vitamin D Release Buffer (50 mL) is an aqueous buffer designed to the fast and efficient release vitamin D and its metabolites from their serum/plasma transporter (vitamin D binding protein) Unlike other release agents this buffer is organic solvent-free and liquid stable For in vitro research use only