Learn More
Sigma Aldrich Fine Chemicals Biosciences Vitamin D2 61919 d3 soluti
Catalog No. 502906027
Encompass
Vitamin D2 is also known as ergocalciferol. It is produced by plants and mushrooms when exposed to sunlight.Vitamin D2nbsp(6 19 19-d3) is a deuterated vitamin D2nbspwherein C-6 and C-19 protons are replaced by deuterium.
Catalog No. 50-290-6027 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 7398471ML
Vitamin D2 is also known as ergocalciferol. It is produced by plants and mushrooms when exposed to sunlight.Vitamin D2nbsp(6 19 19-d3) is a deuterated vitamin D2nbspwherein C-6 and C-19 protons are replaced by deuterium.