Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Vitamin D2, 97+%

Catalog No. AC422630010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC422630010 1 g Glass bottle
AC422630050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC422630010 Supplier Thermo Scientific Chemicals Supplier No. 422630010
  • $117.30
    $98.65 / Each of 1

    Save $18.65

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

Vitamin D2, or Ergocalciferol, CAS # 50-14-6, a fat-soluble hormone that is important for calcium and phosphorus absorption and metabolism. | CAS: 50-14-6 | C28H44O | 396.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Vitamin D2 (ergocalciferol) is a form of vitamin D, a fat-soluble hormone found in food sources
  • Vitamin D is important for calcium and phosphorus homeostasis, and can play a role in many biological processes including cell growth
  • Vitamin D2 has a lower binding affinity than Vitamin D3 for vitamin D binding protein

Applications

  • Research studies show that Vitamin D2 treatment in rats inhibits bladder tumor promotion
  • Studies show that the treatment of lung carcinoma cells with vitamin D2 in vitro resulted in increased ACE2 protein levels
  • Vitamin D2 has been seen to interfere with prion protein oligomerization in vitro
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-14-6
Molecular Formula C28H44O
Molecular Weight (g/mol) 396.66
MDL Number MFCD00166988,MFCD11656674
InChI Key MECHNRXZTMCUDQ-VLOQVYPSSA-N
Synonym ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina
PubChem CID 134129658
IUPAC Name (1S)-3-[2-[(1S,7aR)-1-[(2R,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C

Specifications

Absorbance 460 to 490 (265nm, EtOH 96%)
Melting Point 115.0°C to 119.0°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97.0 to 103% (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 10215
Specific Rotation Condition +82.00 (25.00°C c=3,in acetone)
Specific Rotation + 82.00
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in common organic solvents
Formula Weight 396.65
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Vitamin D2, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air
EINECSNumber 200-014-9
RTECSNumber KE1050000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.