Learn More
Invitrogen™ Vitamin D2/D3 Competitive ELISA Kit
Description
The Vitamin D (VD) ELISA quantitates VD in serum, plasma and other biological fluids. Principle of the method The Competitive mulispecies ELISA research use only kit is designed to quantitatively measure Vitamin D (VD)independent of species . A Vitamin D standard is provided to generate a standard curve for the assay and all samples are read off a user generated standard curve.Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to Vitamin D is added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of Vitamin D in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Specifications
Specifications
| Assay Range | 6.25-400 ng/mL |
| Assay Sensitivity | 3.75 ng/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Cell Lysates, Plasma, Serum, Tissue Homogenates |
| Target Species | All Species |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | calciferol; Cholecalciferol; Ergocalciferol; Vitamin D2; vitamin D3calciferol; Cholecalciferol; Ergocalciferol; Vitamin D2; vitamin D3; |
| Interassay CV | <10% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.