Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vitamin D2 (ergocalciferol) | 50-14-6 | 99.1% | C28H44O | 10 G
SDP

Catalog No. 5000434658
Encompass_Preferred

Vitamin D2 (Ergocalciferol) is derived from plant sources or dietary supplements and can be used as a supplement of Vitamin D. This product is intended for research use only.

  • Derived from plant sources
  • Can be used as a supplement of vitamin D
  • For research use only
  • Molecular weight: 396.65
  • Appearance: solid
  • Color: white to light yellow
  • Structure classification: steroids
  • Initial source: plants other families
  • Endogenous metabolite: human gut microbiota metabolites
  • Shipping at room temperature in continental US
  • Store at -20°C, protected from light, under nitrogen
  • Unstable in solutions; freshly prepared is recommended
  • Solubility (in vitro): DMSO: 31.25 mg/mL (78.78 mM)

Catalog No. 50-004-34658 Supplier Medchemexpress LLC Supplier No. HY7654210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vitamin D2 (Ergocalciferol) is derived from plant sources or dietary supplements and can be used as a supplement of Vitamin D. This product is intended for research use only.

  • Derived from plant sources
  • Can be used as a supplement of vitamin D
  • For research use only
  • Molecular weight: 396.65
  • Appearance: solid
  • Color: white to light yellow
  • Structure classification: steroids
  • Initial source: plants other families
  • Endogenous metabolite: human gut microbiota metabolites
  • Shipping at room temperature in continental US
  • Store at -20°C, protected from light, under nitrogen
  • Unstable in solutions; freshly prepared is recommended
  • Solubility (in vitro): DMSO: 31.25 mg/mL (78.78 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.