Learn More
Biomatik Corporation Vitamin D2 (VD2) CLIA Kit
Supplier:  Biomatik Corporation EKU08649
Encompass_Preferred
Vitamin D2 (VD2) CLIA Kit for detecting VD2 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 1.17-300ng/ml. Sensitivity: 0.45ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: Ergocalciferol; Deltalin; Drisdol; Calcidol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6449