Learn More
Medchemexpress LLC HY-15398 100mg , Vitamin D3 CAS:67-97-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15398100MG
Encompass_Preferred
HY-15398 100mg Vitamin D3 CAS: 67-97-0 Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occuring form of vitamin D. Vitamin D3 induces cell differentiation and prevents proliferation of cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0158