Learn More
Apexbio Technology LLC Vitamin D3(Synonyms: Cholecalciferol, Calciol, Vitamin D₃, Colecalciferol, Activated 7-Dehydrocholesterol), 5g, CAS: 67-97-0.
SDP
Catalog No. 5000060569
Encompass_Preferred
Vitamin D3 (Cholecalciferol CAS 67-97-0) is an endogenous fat-soluble vitamin that serves as a precursor requiring metabolic conversion to its biologically active metabolite Following hydroxylation the active form exerts its function predominantly by binding the nuclear vitamin D receptor (VDR) modulating gene expression pathways involved in cellular differentiation and proliferation Research indicates that activated Cholecalciferol promotes cellular differentiation and exhibits anti-proliferative effects against various cancer cell types Consequently Vitamin D3 is frequently utilized in biomedical research studying mechanisms underlying tumor development and differentiation pathways
Catalog No. 50-000-60569 Supplier Apexbio Technology LLC Supplier No. B20655000
Add to Cart
Add to Cart
Vitamin D3 (Cholecalciferol CAS 67-97-0) is an endogenous fat-soluble vitamin that serves as a precursor requiring metabolic conversion to its biologically active metabolite Following hydroxylation the active form exerts its function predominantly by binding the nuclear vitamin D receptor (VDR) modulating gene expression pathways involved in cellular differentiation and proliferation Research indicates that activated Cholecalciferol promotes cellular differentiation and exhibits anti-proliferative effects against various cancer cell types Consequently Vitamin D3 is frequently utilized in biomedical research studying mechanisms underlying tumor development and differentiation pathways