Learn More
Medchemexpress LLC Vitamin D3 | 67-97-0 | C27H44O | 5 G
SDP
Catalog No. 5000376515
Encompass_Preferred
Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occurring form of vitamin D. It induces cell differentiation and prevents the proliferation of cancer cells, playing a crucial role in various biological activities for research use.
  • Naturally occurring form of vitamin D.
  • Induces cell differentiation and prevents proliferation of cancer cells.
  • Undergoes two hydroxylation processes to become biologically active in vivo.
  • Active form is 1,25-dihydroxycholecalciferol (calcitriol).
  • Exhibits anti-proliferative effects in various cell lines in a dose- and time-dependent manner.
  • Increases nuclear VDR staining and produces local VDR activation in IK cells.
  • Potentiates CCl4 toxicity in the liver and increases renal calcium levels in mice in vivo.
Catalog No. 50-003-76515 Supplier Medchemexpress LLC Supplier No. HY153985G
Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occurring form of vitamin D. It induces cell differentiation and prevents the proliferation of cancer cells, playing a crucial role in various biological activities for research use.
  • Naturally occurring form of vitamin D.
  • Induces cell differentiation and prevents proliferation of cancer cells.
  • Undergoes two hydroxylation processes to become biologically active in vivo.
  • Active form is 1,25-dihydroxycholecalciferol (calcitriol).
  • Exhibits anti-proliferative effects in various cell lines in a dose- and time-dependent manner.
  • Increases nuclear VDR staining and produces local VDR activation in IK cells.
  • Potentiates CCl4 toxicity in the liver and increases renal calcium levels in mice in vivo.