Learn More
Medchemexpress LLC Vitamin D3-d3 | 80666-48-4 | C27H41D3O | 1 MG
SDP
Catalog No. 5000376499
Encompass_Preferred
Vitamin D3-d3 is the deuterium labeled Vitamin D3. Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occurring form of vitamin D. It induces cell differentiation and prevents proliferation of cancer cells.
  • Deuterium labeled vitamin D3
  • Naturally occurring form of vitamin D
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-76499 Supplier Medchemexpress LLC Supplier No. HY15398S31MG
Vitamin D3-d3 is the deuterium labeled Vitamin D3. Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occurring form of vitamin D. It induces cell differentiation and prevents proliferation of cancer cells.
  • Deuterium labeled vitamin D3
  • Naturally occurring form of vitamin D
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS