Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vitamin D3-d3 | 80666-48-4 | C27H41D3O | 100 UG
SDP

Catalog No. 5000376342
Encompass_Preferred

Vitamin D3-d3 is a deuterium labeled Vitamin D3. This naturally occurring form of vitamin D induces cell differentiation and prevents the proliferation of cancer cells.

  • Deuterium labeled
  • Naturally occurring form of vitamin D
  • Solid, white to off-white appearance
  • Purity: 98.45%
  • Storage: -80°C, protect from light, stored under nitrogen
  • Soluble in ethanol (1 mg/mL)
  • Shipping with dry ice
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Catalog No. 50-003-76342 Supplier Medchemexpress LLC Supplier No. HY15398S3100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vitamin D3-d3 is a deuterium labeled Vitamin D3. This naturally occurring form of vitamin D induces cell differentiation and prevents the proliferation of cancer cells.

  • Deuterium labeled
  • Naturally occurring form of vitamin D
  • Solid, white to off-white appearance
  • Purity: 98.45%
  • Storage: -80°C, protect from light, stored under nitrogen
  • Soluble in ethanol (1 mg/mL)
  • Shipping with dry ice
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.