Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D3, MP Biomedicals™

Catalog No. MP021012242 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021012242 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP021012242 Supplier MP Biomedicals, Inc Supplier No. 02101224.2
Add to Cart
Add to Cart

Is the naturally occurring form of vitamin D3. Vitamin D3, MP Biomedicals mediates intestinal calcium absorption, bone metabolism, and probably muscle activity. It functions as a hormone precursor and acts through a receptor that is a member of the ligand

It acts as a hormone precursor since it requires two stages of metabolism: first to 25-hydroxycholecalciferol; then to 1,25-dihydroxycholecalciferol.

  • Modulates the proliferation and differentiation of both normal and cancer cells
  • Has antiproliferative and antimetastatic effects on breast, colon, and prostate cancer cells
  • Activated vitamin D receptors in intestine and bone maintain calcium absorbance and homeostasis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-97-0
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.648
InChI Key QYSXJUFSXHHAJI-YRZJJWOYSA-N
Synonym vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum
PubChem CID 5280795
ChEBI CHEBI:28940
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C

Specifications

Melting Point 82°C to 92°C
Quantity 250 mg
Solubility Information Soluble in ethanol, acetone, ether, chloroform; insoluble in water. Practically insoluble in water: soluble in the usual organic solvents; slightly soluble in vegetable oils.
Formula Weight 384.65
Physical Form Powder
Recommended Storage Store at 4 °C, desiccate.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.