Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D3, MP Biomedicals™

Catalog No. ICN10122405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10122405 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10122405 Supplier MP Biomedicals, Inc Supplier No. 0210122405
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-97-0
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.648
MDL Number MFCD00078131
InChI Key QYSXJUFSXHHAJI-YRZJJWOYSA-N
Synonym vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum
PubChem CID 5280795
ChEBI CHEBI:28940
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C

Specifications

Color White
Quantity 5 g
Formula Weight 384.65
Physical Form Powder
Chemical Name or Material Vitamin D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.