Learn More
Medchemexpress LLC Vitamin D3 octanoate | 927822-16-0 | 1 MG
SDP
Catalog No. 5000419732
Encompass_Preferred
Vitamin D3 octanoate is an octanoate ester of vitamin D3. Vitamin D3 (Cholecalciferol) is a naturally occurring form of vitamin D that induces cell differentiation and prevents the proliferation of cancer cells. This product is for research use only.
  • Octanoate ester of vitamin D3
  • Naturally occurring form of vitamin D
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
Catalog No. 50-004-19732 Supplier Medchemexpress LLC Supplier No. HYN83571MG
Vitamin D3 octanoate is an octanoate ester of vitamin D3. Vitamin D3 (Cholecalciferol) is a naturally occurring form of vitamin D that induces cell differentiation and prevents the proliferation of cancer cells. This product is for research use only.
  • Octanoate ester of vitamin D3
  • Naturally occurring form of vitamin D
  • Induces cell differentiation
  • Prevents proliferation of cancer cells