Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vitamin D3 octanoate | 927822-16-0 | 10 MG
SDP

Catalog No. 5000417504
Encompass_Preferred

Vitamin D3 octanoate is an octanoate ester of vitamin D3. Vitamin D3 (Cholecalciferol) is a naturally occurring form of Vitamin D. Vitamin D3 induces cell differentiation and prevents proliferation of cancer cells.

  • Octanoate ester of vitamin D3
  • Naturally occurring form of vitamin D
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
  • Colorless to light yellow oil appearance
  • Purity of 90.0%
  • Requires storage at -80°C, protected from light and under nitrogen
  • Solubility in DMSO at ≥ 100 mg/mL

Catalog No. 50-004-17504 Supplier Medchemexpress LLC Supplier No. HYN835710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vitamin D3 octanoate is an octanoate ester of vitamin D3. Vitamin D3 (Cholecalciferol) is a naturally occurring form of Vitamin D. Vitamin D3 induces cell differentiation and prevents proliferation of cancer cells.

  • Octanoate ester of vitamin D3
  • Naturally occurring form of vitamin D
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
  • Colorless to light yellow oil appearance
  • Purity of 90.0%
  • Requires storage at -80°C, protected from light and under nitrogen
  • Solubility in DMSO at ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.