Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitamin D3 Powder, With Corn Starch, MP Biomedicals™

Catalog No. ICN10328080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10328080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10328080 Supplier MP Biomedicals, Inc Supplier No. 0210328080
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-97-0
Molecular Formula C27H44O
Molecular Weight (g/mol) 384.648
InChI Key QYSXJUFSXHHAJI-YRZJJWOYSA-N
Synonym vitamin d3, cholecalciferol, calciol, colecalciferol, oleovitamin d3, arachitol, activated 7-dehydrocholesterol, vitamin d, colecalcipherol, colecalciferolum
PubChem CID 5280795
ChEBI CHEBI:28940
IUPAC Name (1S,3Z)-3-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexan-1-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCCC2=CC=C3CC(CCC3=C)O)C

Specifications

Quantity 100 g
Formula Weight 384.6
Physical Form Solid
Chemical Name or Material Vitamin D3 Powder
Recommended Storage Store at 2° to 8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.