Learn More
Medchemexpress LLC Vitamin D3 (Standard) | 67-97-0 | C27H44O | 25 MG
SDP
Catalog No. 5000375202
Encompass_Preferred
Vitamin D3 (Standard) is the analytical standard of Vitamin D3, intended for research and analytical applications. This naturally occurring form of vitamin D induces cell differentiation and prevents the proliferation of cancer cells. It serves as a reference standard for qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-003-75202 Supplier Medchemexpress LLC Supplier No. 25MGHY15398R
Vitamin D3 (Standard) is the analytical standard of Vitamin D3, intended for research and analytical applications. This naturally occurring form of vitamin D induces cell differentiation and prevents the proliferation of cancer cells. It serves as a reference standard for qualitative, quantitative, and methodological research experiments, including HPLC, GC, and MS.
  • Induces cell differentiation
  • Prevents proliferation of cancer cells
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)