Learn More
Medchemexpress LLC Vitamin D3 (Standard) | 67-97-0 | C27H44O | 50 MG
SDP
Catalog No. 5000376111
Encompass_Preferred
Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occurring form of vitamin D and serves as an analytical standard. This product is intended for research and analytical applications, inducing cell differentiation and preventing the proliferation of cancer cells.
  • Analytical standard for Vitamin D3
  • Intended for research and analytical applications
  • Induces cell differentiation and prevents proliferation of cancer cells
  • Commonly used in qualitative, quantitative, and methodological research
  • Suitable for HPLC, GC, and MS applications
  • Naturally occurring form of vitamin D
Catalog No. 50-003-76111 Supplier Medchemexpress LLC Supplier No. HY15398R50MG
Vitamin D3 (Cholecalciferol; Colecalciferol) is a naturally occurring form of vitamin D and serves as an analytical standard. This product is intended for research and analytical applications, inducing cell differentiation and preventing the proliferation of cancer cells.
  • Analytical standard for Vitamin D3
  • Intended for research and analytical applications
  • Induces cell differentiation and prevents proliferation of cancer cells
  • Commonly used in qualitative, quantitative, and methodological research
  • Suitable for HPLC, GC, and MS applications
  • Naturally occurring form of vitamin D