Learn More
Medchemexpress LLC Vitamin D4 (22-Dihydroergocalciferol) | 511-28-4 | 99.5% | C28H46O | 25 MG
SDP
Catalog No. 5000357979
Encompass_Preferred
Vitamin D4 (22-Dihydroergocalciferol) is a Vitamin D derived from fungi, with 22,23-dihydroergosterol as its precursor. This product is intended for research use only and is not sold for patient use.
  • Derived from fungi
  • Precursor is 22,23-dihydroergosterol
  • For research use only
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO at 25 mg/mL
Catalog No. 50-003-57979 Supplier Medchemexpress LLC Supplier No. 25MGHY75958
Vitamin D4 (22-Dihydroergocalciferol) is a Vitamin D derived from fungi, with 22,23-dihydroergosterol as its precursor. This product is intended for research use only and is not sold for patient use.
  • Derived from fungi
  • Precursor is 22,23-dihydroergosterol
  • For research use only
  • Appears as an off-white to light yellow solid
  • Soluble in DMSO at 25 mg/mL