Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Vitamin D4 (22-dihydroergocalciferol) | 511-28-4 | 99.5% | C28H46O | 50 MG
SDP

Catalog No. 5000433682
Encompass_Preferred

Vitamin D4 (22-Dihydroergocalciferol) is a vitamin D derived from fungi. The precursor of vitamin D4 is 22,23-dihydroergosterol. It is for research use only and not sold to patients.

  • Appearance: Solid
  • Color: Off-white to light yellow
  • Molecular weight: 398.66
  • Solubility: 25 mg/mL in DMSO

Catalog No. 50-004-33682 Supplier Medchemexpress LLC Supplier No. HY7595850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Vitamin D4 (22-Dihydroergocalciferol) is a vitamin D derived from fungi. The precursor of vitamin D4 is 22,23-dihydroergosterol. It is for research use only and not sold to patients.

  • Appearance: Solid
  • Color: Off-white to light yellow
  • Molecular weight: 398.66
  • Solubility: 25 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.