Learn More
Selleck Chemical LLC Vitamin E S4686-500mg
SDP
Catalog No. 503112007
Encompass_Preferred
Vitamin E (D-alpha-Tocopherol) is a fat-soluble vitamin with potent antioxidant properties It is a potent peroxyl radical scavenger and inhibits noncompetitively cyclooxygenase activity in many tissues also inhibits angiogenesis and tumor dormancy through suppressing vascular endothelial growth factor (VEGF) gene transcription
Catalog No. 50-311-2007 Supplier Selleck Chemical LLC Supplier No. S4686500MG
Vitamin E (D-alpha-Tocopherol) is a fat-soluble vitamin with potent antioxidant properties It is a potent peroxyl radical scavenger and inhibits noncompetitively cyclooxygenase activity in many tissues also inhibits angiogenesis and tumor dormancy through suppressing vascular endothelial growth factor (VEGF) gene transcription