Learn More
Selleck Chemical LLC Vitamin E acetate-E0191-1G
SDP
Catalog No. 503137576
Encompass_Preferred
DL- -Tocopherol acetate (DL-Alpha-tocopherol acetate Vitamin E acetate) a vitamin E derivative is used as a vitamin E supplement in poultry feeds
Catalog No. 50-313-7576 Supplier Selleck Chemical LLC Supplier No. E01911G
DL- -Tocopherol acetate (DL-Alpha-tocopherol acetate Vitamin E acetate) a vitamin E derivative is used as a vitamin E supplement in poultry feeds