Learn More
U.S. Pharmacopeia Alpha Tocopherol, 10191-41-0, MFCD00006848, 250 mg
Supplier:  U.S. Pharmacopeia 1667600
Encompass
Empirical Formula (Hill Notation): C29H50O2, Molecular Weight: 430.71, Synonym: (±)-α-Tocopherol, DL-all-rac-α-Tocopherol, Vitamin E.
Catalog No. NC9154894