Learn More
Abcam Vitamin E (DL-alpha-Tocopherol), Fat-soluble antio x idant, 25G
Catalog No. 502706143
Encompass_Preferred
MW 430.7 Da, Purity >98%. Fat-soluble antioxidant. Peroxyl radical scavenger that protects polyunsaturated fatty acids in membranes and lipoproteins. Anticarcinogen.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6143 Supplier Abcam Supplier No. AB14296825G
MW 430.7 Da, Purity >98%. Fat-soluble antioxidant. Peroxyl radical scavenger that protects polyunsaturated fatty acids in membranes and lipoproteins. Anticarcinogen.
The product is subject to the following: Abcam Restricted Use Statement