Learn More
ANTARES Vitamin E TPGS | 9002-96-4 | 50 g SAMPLE
Catalog No. NC3376612
Encompass_Preferred
Antares Vitamin E TPGS (d-alpha tocopheryl polyethylene glycol 1000 succinate, TPGS, Tocophersolan, CAS: 9002-96-4) is a powerful tool in the formulation of lipophilic and poorly soluble compounds and in enhancing their absorption and bioavailability. TPGS has a clinically proven record of safety and efficacy in pharmaceutical, nutrition, food and beverage, personal care and animal health and nutrition products. Its unique properties, which include inhibition of the P-Glycoprotein efflux system and synergies with active pharmaceutical ingredients (APIs) and prodrug formulations, support emerging novel applications in drug delivery. Antares Health Products, Inc. is a leading supplier of TPGS. Its team combines diverse experience and works in close partnership with its customers in support of product innovation
Catalog No. NC3376612 Supplier ANTARES Supplier No. TG0150NF
Antares Vitamin E TPGS (d-alpha tocopheryl polyethylene glycol 1000 succinate, TPGS, Tocophersolan, CAS: 9002-96-4) is a powerful tool in the formulation of lipophilic and poorly soluble compounds and in enhancing their absorption and bioavailability. TPGS has a clinically proven record of safety and efficacy in pharmaceutical, nutrition, food and beverage, personal care and animal health and nutrition products. Its unique properties, which include inhibition of the P-Glycoprotein efflux system and synergies with active pharmaceutical ingredients (APIs) and prodrug formulations, support emerging novel applications in drug delivery. Antares Health Products, Inc. is a leading supplier of TPGS. Its team combines diverse experience and works in close partnership with its customers in support of product innovation