Learn More
Medchemexpress LLC Vitamin K | 12001-79-5 | 98.7% | 25 MG
SDP
Catalog No. 5000411273
Encompass_Preferred
Vitamin K, the blood-clotting vitamin, is crucial for many body proteins, including coagulation factors, osteocalcin, and matrix-Gla protein. It supports nervous system function, activates specific proteins (Gas6, S), and aids brain sphingolipid synthesis. Both Phylloquinone (K1) and Menaquinones (K2) are vital for γ-glutamyl carboxylation. Supplementation may improve insulin sensitivity, glucose tolerance, prevent insulin resistance, reduce type 2 diabetes risk, and support bone health.
  • Supports proper function of numerous body proteins
  • Essential for nervous system health and brain sphingolipid synthesis
  • Contributes to biological activation of specific proteins
  • Vital for γ-glutamyl carboxylation of vitamin K-dependent proteins
  • May improve metabolic health, including insulin sensitivity and glucose tolerance
  • Supports bone health
Catalog No. 50-004-11273 Supplier Medchemexpress LLC Supplier No. 25MGHYB2172
Vitamin K, the blood-clotting vitamin, is crucial for many body proteins, including coagulation factors, osteocalcin, and matrix-Gla protein. It supports nervous system function, activates specific proteins (Gas6, S), and aids brain sphingolipid synthesis. Both Phylloquinone (K1) and Menaquinones (K2) are vital for γ-glutamyl carboxylation. Supplementation may improve insulin sensitivity, glucose tolerance, prevent insulin resistance, reduce type 2 diabetes risk, and support bone health.
  • Supports proper function of numerous body proteins
  • Essential for nervous system health and brain sphingolipid synthesis
  • Contributes to biological activation of specific proteins
  • Vital for γ-glutamyl carboxylation of vitamin K-dependent proteins
  • May improve metabolic health, including insulin sensitivity and glucose tolerance
  • Supports bone health