Learn More
TARGETMOL CHEMICALS INC Vitamin K 25MG
SDP
Catalog No. 502876010
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. Vitamin K (Kephton) has blood coagulation action and is important for coagulation factors, matrix-Gla protein, and osteocalcin. Purity 99.79%
Catalog No. 50-287-6010 Supplier TARGETMOL CHEMICALS INC Supplier No. T1960325MG
Also available in 5 mg, 10 mg, 50 mg and bulk. Please contact Fisher for quotes. Vitamin K (Kephton) has blood coagulation action and is important for coagulation factors, matrix-Gla protein, and osteocalcin. Purity 99.79%