Learn More
Creative Biomart Coagulation factor X human
Catalog No. 502084112
Encompass_Preferred
The product, is a plasma-derived human blood coagulation factor. Coagulation Factor X is a vitamin K-dependent, liver-produced serine protease that serves as the first enzyme in the coagulation cascade to form fibrin. It is a two-chain glycoprotein with the molecular weight of approximately 59 kDa. While Factor X normally circulates in the plasma as inactive molecules, the activation of Factor X is involved in both the intrinsic and extrinsic coagulation pathways.
Catalog No. 50-208-4112 Supplier Creative Biomart Supplier No. THP0061
The product, is a plasma-derived human blood coagulation factor. Coagulation Factor X is a vitamin K-dependent, liver-produced serine protease that serves as the first enzyme in the coagulation cascade to form fibrin. It is a two-chain glycoprotein with the molecular weight of approximately 59 kDa. While Factor X normally circulates in the plasma as inactive molecules, the activation of Factor X is involved in both the intrinsic and extrinsic coagulation pathways.