Learn More
Creative Biomart Protein S human
Catalog No. 502084140
Encompass_Preferred
The product is a vitamin K-dependent plasma glycoprotein which has antcoagulant properties. It serves as a negative feedback mechanism in the coagulation cascade.
Catalog No. 50-208-4140 Supplier Creative Biomart Supplier No. THP0089
The product is a vitamin K-dependent plasma glycoprotein which has antcoagulant properties. It serves as a negative feedback mechanism in the coagulation cascade.