Learn More
Medchemexpress LLC HY-N0684 1g Medchemexpress, Vitamin K1 CAS:84-80-0 Purity:>98%
SDP
Catalog No. 501933676
Encompass_Preferred
Medchemexpress, HY-N0684 1g Vitamin K1 CAS:84-80-0 Vitamin K1 a fat-soluble, naturally occurring vitamin required for blood coagulation and bone and vascular metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3676 Supplier Medchemexpress LLC Supplier No. HYN06841G
Medchemexpress, HY-N0684 1g Vitamin K1 CAS:84-80-0 Vitamin K1 a fat-soluble, naturally occurring vitamin required for blood coagulation and bone and vascular metabolism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.