Learn More
Cayman Chemical Vitamin K1 2,3-epoxide, Purity: ≥95%, CAS: 25486-55-9, 1 mg
SDP
Catalog No. 502971805
Encompass_Preferred
The inactive metabolite of vitamin K1 formed during blood clotting that is rapidly reduced back to vitamin K1 by a microsomal epoxide reductase
Catalog No. 50-297-1805 Supplier Cayman Chemical Supplier No. 160171 MG
The inactive metabolite of vitamin K1 formed during blood clotting that is rapidly reduced back to vitamin K1 by a microsomal epoxide reductase