Learn More
Medchemexpress LLC Vitamin K1 2,3-epoxide | 25486-55-9 | 99.99% | 466.70 | 1 MG
SDP
Catalog No. 5000435958
Encompass_Preferred
Vitamin K1 2,3-epoxide is an inactive metabolite form of Vitamin K1, which is reduced to active vitamin by microsomal epoxide reductase in the vitamin K epoxide cycle. This compound is involved in blood clotting.
  • Inactive metabolite form of Vitamin K1.
  • Reduced to active vitamin by microsomal epoxide reductase.
  • Involved in blood clotting.
Catalog No. 50-004-35958 Supplier Medchemexpress LLC Supplier No. HY1131641MG
Vitamin K1 2,3-epoxide is an inactive metabolite form of Vitamin K1, which is reduced to active vitamin by microsomal epoxide reductase in the vitamin K epoxide cycle. This compound is involved in blood clotting.
  • Inactive metabolite form of Vitamin K1.
  • Reduced to active vitamin by microsomal epoxide reductase.
  • Involved in blood clotting.