Learn More
Cayman Chemical VItamIn K1 2 3-epoxIde 500ug
SDP
Catalog No. 502971804
Encompass_Preferred
The inactive metabolite of vitamin K1 formed during blood clotting that is rapidly reduced back to vitamin K1 by a microsomal epoxide reductase
Catalog No. 50-297-1804 Supplier Cayman Chemical Supplier No. 16017500 UG
The inactive metabolite of vitamin K1 formed during blood clotting that is rapidly reduced back to vitamin K1 by a microsomal epoxide reductase