Learn More
Selleck Chemical LLC Vitamin K1 S4698-25mg
SDP
Catalog No. 503110117
Encompass_Preferred
Vitamin K1 (Phyllohydroquinone Phylloquinone Phytomenadione Phytonadione) made by plants is a major type of dietary vitamin K which is well-known for its role in blood clotting Vitamin K1 is directly involved in photosynthesis
Catalog No. 50-311-0117 Supplier Selleck Chemical LLC Supplier No. S469825MG
Vitamin K1 (Phyllohydroquinone Phylloquinone Phytomenadione Phytonadione) made by plants is a major type of dietary vitamin K which is well-known for its role in blood clotting Vitamin K1 is directly involved in photosynthesis