Learn More
Sigma Aldrich Fine Chemicals Biosciences Vitamin K1 4a56788a 13C6 91MG
Catalog No. 502907918
Encompass
Vitamin K1 also known as phylloquinone is a fat-soluble vitamin found in algae nbspcyanobacteria and plants. It is involved in the synthesis of some important proteins having applications in blood clotting and bone metabolism.Vitamin K1-4a 5 6 7 8 8a-13C6 is an isotope of vitamin K1 wherein C-4a C-5 C-6 C-7 C-8 C-8a carbons are replaced by 13C6 carbon.
Catalog No. 50-290-7918 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 8098881MG
Vitamin K1 also known as phylloquinone is a fat-soluble vitamin found in algae nbspcyanobacteria and plants. It is involved in the synthesis of some important proteins having applications in blood clotting and bone metabolism.Vitamin K1-4a 5 6 7 8 8a-13C6 is an isotope of vitamin K1 wherein C-4a C-5 C-6 C-7 C-8 C-8a carbons are replaced by 13C6 carbon.