Learn More
Enzo Life Sciences Vitamin K1 (5g). CAS: 84-80-0
SDP
Supplier:  Enzo Life Sciences ALX460027G005
Encompass_Preferred
Alternative name: Phytomenadione, Phylloquinone, 2-Methyl-3-phytyl-1,4-naphthoquinone. Purity: ≥98% (TLC). Solubility: Soluble in acetone, 100% ethanol, chloroform, ether, hexane and vegetable oils; slightly soluble in methanol; insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-9291