Learn More
Medchemexpress LLC Vitamin K1-d4 | 5172-18-9 | 454.72 | 1 MG
SDP
Catalog No. 5000416137
Encompass_Preferred
Vitamin K1-d4 is the deuterium labeled Vitamin K1, a naturally occurring vitamin essential for blood coagulation, bone, and vascular metabolism. This compound is valuable as a tracer and as an internal standard for quantitative analysis in various analytical techniques. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules to serve as tracers for quantitation during drug development, potentially influencing pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-16137 Supplier Medchemexpress LLC Supplier No. HYN0684S11MG
Vitamin K1-d4 is the deuterium labeled Vitamin K1, a naturally occurring vitamin essential for blood coagulation, bone, and vascular metabolism. This compound is valuable as a tracer and as an internal standard for quantitative analysis in various analytical techniques. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules to serve as tracers for quantitation during drug development, potentially influencing pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.