Learn More
Medchemexpress LLC Vitamin K1 (Standard) | 84-80-0 | MFCD00214063 | 450.70 | 50 MG
SDP
Catalog No. 5000415443
Encompass_Preferred
Vitamin K1 (Standard) is the analytical standard of Vitamin K1, intended for research and analytical applications. It is a naturally occurring vitamin required for blood coagulation and bone and vascular metabolism.
  • Analytical standard, reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Endogenous metabolite.
  • Initial source: Plants, other families, Human gut microbiota, Microorganisms.
  • Structure classification: Quinones, Naphthalene quinones.
Catalog No. 50-004-15443 Supplier Medchemexpress LLC Supplier No. HYN0684R50MG
Vitamin K1 (Standard) is the analytical standard of Vitamin K1, intended for research and analytical applications. It is a naturally occurring vitamin required for blood coagulation and bone and vascular metabolism.
  • Analytical standard, reference standard supplied assay.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Endogenous metabolite.
  • Initial source: Plants, other families, Human gut microbiota, Microorganisms.
  • Structure classification: Quinones, Naphthalene quinones.