Learn More
TARGETMOL CHEMICALS INC VITAMIN K2 25MG
SDP
Catalog No. 502580876
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Vitamin K2 (Menatetrenone) is a menaquinone compound and form of vitamin K2 with potential antineoplastic activity. Menatetrenone may act by modulating the signaling of certain tyrosine kinases thereby affecting several transcription factors including c-myc and c-fos. This agent inhibits tumor cell growth by inducing apoptosis and cell cycle arrest. purity: 99%
Catalog No. 50-258-0876 Supplier TARGETMOL CHEMICALS INC Supplier No. T233625MG
Also available in 5 mg 10 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Vitamin K2 (Menatetrenone) is a menaquinone compound and form of vitamin K2 with potential antineoplastic activity. Menatetrenone may act by modulating the signaling of certain tyrosine kinases thereby affecting several transcription factors including c-myc and c-fos. This agent inhibits tumor cell growth by inducing apoptosis and cell cycle arrest. purity: 99%